missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylfuran, 98+%, Thermo Scientific Chemicals

Catalog Number p-7022646
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
3208-16-0
C6H8O
96.13
MFCD00003259
HLPIHRDZBHXTFJ-UHFFFAOYSA-N
furan, 2-ethyl, alpha-ethylfuran, 2-ethyloxole, 2-ethyl furan, ccris 4387, unii-t8o6j71t9o, fema no. 3673, furan, .alpha.-ethyl, a-ethylfuran, 2-ethyl furane
18554
2-ethylfuran
CCC1=CC=CO1
This item is not returnable. View return policy
3208-16-0
C6H8O
96.13
MFCD00003259
HLPIHRDZBHXTFJ-UHFFFAOYSA-N
furan, 2-ethyl, alpha-ethylfuran, 2-ethyloxole, 2-ethyl furan, ccris 4387, unii-t8o6j71t9o, fema no. 3673, furan, .alpha.-ethyl, a-ethylfuran, 2-ethyl furane
18554
2-ethylfuran
CCC1=CC=CO1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.912
Boiling Point 92°C to 93°C
Flash Point −2°C (28°F)
Refractive Index 1.44
Quantity 5 g
UN Number UN3271
Beilstein 105401
Sensitivity Air Sensitive
Formula Weight 96.13
Percent Purity ≥98%
Chemical Name or Material 2-Ethylfuran
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'