Learn More
2-(Ethylamino)-1-phenylpropan-1-ol Hydrochloride, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 2-(Ethylamino)-1-phenylpropan-1-ol hydrochloride |
| CAS | 63401-08-1 |
| Analyte or Component Names | 2-(Ethylamino)-1-phenylpropan-1-ol hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H17 N O . Cl H |
| Synonym | Benzenemethanol, α-[1-(ethylamino)ethyl]-, hydrochloride (1:1), Benzenemethanol, α-[1-(ethylamino)ethyl]-, hydrochloride (9CI), Benzyl alcohol, α-(1-ethylaminoethyl)-, -HCl (4CI), RMI 8205A, 2-(Ethylamino)-1-phenylpropan-1-ol Hydrochloride, (±)-N-Ethylcathinone ephedrine metabolite hydrochloride |
| InChI Formula | InChI=1S/C11H17NO.ClH/c1-3-12-9(2)11(13)10-7-5-4-6-8-10;/h4-9,11-13H,3H2,1-2H3;1H |
| Solution Type | Neat |
| SMILES | Cl.CCNC(C)C(O)c1ccccc1 |
| IUPAC Name | 2-(ethylamino)-1-phenylpropan-1-ol;hydrochloride |
| Show More |