Learn More
2-(Ethylamino)-1-phenylpropan-1-ol Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal
Product Code. 30458530
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. 30458530
Brand: LoGiCal LGCAMP0718.0101
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(Ethylamino)-1-phenylpropan-1-ol hydrochloride
CAS 63401-08-1
Analyte or Component Names 2-(Ethylamino)-1-phenylpropan-1-ol hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C11 H17 N O . Cl H
Concentration 1 mg/mL
Synonym Benzenemethanol, α-[1-(ethylamino)ethyl]-, hydrochloride (1:1), Benzenemethanol, α-[1-(ethylamino)ethyl]-, hydrochloride (9CI), Benzyl alcohol, α-(1-ethylaminoethyl)-, -HCl (4CI), RMI 8205A, 2-(Ethylamino)-1-phenylpropan-1-ol Hydrochloride, (±)-N-Ethylcathinone ephedrine metabolite hydrochloride
InChI Formula InChI=1S/C11H17NO.ClH/c1-3-12-9(2)11(13)10-7-5-4-6-8-10;/h4-9,11-13H,3H2,1-2H3;1H
Solution Type Single Solution
SMILES Cl.CCNC(C)C(O)c1ccccc1
IUPAC Name 2-(ethylamino)-1-phenylpropan-1-ol;hydrochloride
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 215.72
Quantity 1 mL
Formula Weight 215.1077 g/mol