missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxy-3,5-difluorobenzaldehyde, 97%, Thermo Scientific™

Catalog Number 15455007
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
883535-74-8
C9H8F2O2
186.158
MFCD06247321
RQGAAZRZCQTFPA-UHFFFAOYSA-N
3,5-difluoro-2-ethoxybenzaldehyde, 2-ethoxy-3,5-difluorobenzaldehyde, jrd
46737626
2-ethoxy-3,5-difluorobenzaldehyde
CCOC1=C(C=C(C=C1C=O)F)F
This item is not returnable. View return policy
883535-74-8
C9H8F2O2
186.158
MFCD06247321
RQGAAZRZCQTFPA-UHFFFAOYSA-N
3,5-difluoro-2-ethoxybenzaldehyde, 2-ethoxy-3,5-difluorobenzaldehyde, jrd
46737626
2-ethoxy-3,5-difluorobenzaldehyde
CCOC1=C(C=C(C=C1C=O)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 186.16
Percent Purity 97%
Chemical Name or Material 2-Ethoxy-3,5-difluorobenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'