Learn More
2-Ethoxy-1-naphthaldehyde, 98%, Thermo Scientific™
Product Code. 11426986
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426986 5 g 5g
11416986 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11426986 Supplier Thermo Scientific Alfa Aesar Supplier No. B25306.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 19523-57-0
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.24
MDL Number MFCD00046427
InChI Key IMNKQTWVJHODOS-UHFFFAOYSA-N
Synonym 2-ethoxy-1-naphthaldehyde, 1-naphthalenecarboxaldehyde, 2-ethoxy, 2-ethoxy-1-naphthalenecarboxaldehyde, 1-naphthaldehyde, 2-ethoxy, 1-naphthalenecarboxaldehyde,2-ethoxy, 2-ethoxynaphthalenecarbaldehyde, 2-athoxy-naphthal, acmc-1bs4w, 2-ethoxy-1-naphthalenecarbaldehyde, 2-ethoxy-naphthalene-1-carbaldehyde
PubChem CID 88099
IUPAC Name 2-ethoxynaphthalene-1-carbaldehyde
SMILES CCOC1=CC=C2C=CC=CC2=C1C=O
Specifications
Melting Point 111°C to 113°C
Quantity 5 g
Beilstein 1073578
Sensitivity Air Sensitive
Formula Weight 200.24
Percent Purity 98%
Chemical Name or Material 2-Ethoxy-1-naphthaldehyde