Learn More
2-(Dimethylcarbamoyl)pyridine-5-boronic acid pinacol ester, 96%
Product Code. p-7036337
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15471396
Brand: Thermo Scientific Alfa Aesar H54614.MD
Request Bulk or Custom Format
CAS: 1006876-27-2 | C14H21BN2O3 | 276.143 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1006876-27-2
Molecular Formula C14H21BN2O3
Molecular Weight (g/mol) 276.143
MDL Number MFCD08458484
InChI Key RNCGUUHRBRONRG-UHFFFAOYSA-N
Synonym n,n-dimethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl picolinamide, 6-dimethylcarbamoyl pyridin-3-ylboronic acid pinacol ester, n,n-dimethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-carboxamide, 6-dimethylcarbamoyl pyridine-3-boronic acid pinacol ester, 2-dimethylcarbamoyl pyridine-5-boronic acid pinacol ester, n,n-dimethyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-carboxamide, 2-n,n-dimthylaminocarbonyl pyridine-5-boronic acid pinacol ester, n,n-dimethyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl picolinamide, 2-pyridinecarboxamide, n,n-dimethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, n,n-dimethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinecarboxamide
PubChem CID 57416491
IUPAC Name N,N-dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)C(=O)N(C)C
Specifications
Quantity 250 mg
Formula Weight 276.14
Percent Purity 96%
Chemical Name or Material 2-(Dimethylcarbamoyl)pyridine-5-boronic acid pinacol ester