Learn More
2-(Dimethylamino)-ethanol, Sigma-Aldrich™
Product Code. 15894111
Change view
Click to view available options
Quantity:
100 mL
1 L
25 kg
Unit Size:
1 L
100mL
25kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15894111 100 mL 100mL
15804121 1 L 1 L
15814121 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15894111 Supplier Sigma Aldrich Supplier No. 8032370100
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 108-01-0
Grade Research
Molecular Formula (CH3)2NCH2CH2OH
Molecular Weight (g/mol) 89.14
Specifications
Melting Point -59°C
Density 0.89 g/cm3 at 20°C
MDL Number MFCD00002846
Quantity 100 mL
Synonym 2-(Dimethylamino)-ethanol, N,N-Dimethylethanolamine, 2-(Dimethylamino)-ethanol