Learn More
2-Diethylaminoethyl Hexanoate, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Diethylaminoethyl Hexanoate |
| CAS | 10369-83-2 |
| Molecular Formula | C12 H25 N O2 |
| Quantity | 50 mg |
| Synonym | Hexanoic acid, 2-(diethylamino)ethyl ester, 2-(Diethylamino)ethyl caproate, 2-Diethylaminoethyl hexanoate, DA 6, Diethyl aminoethyl hexanoate, HKL 4 |
| InChI Formula | InChI=1S/C12H25NO2/c1-4-7-8-9-12(14)15-11-10-13(5-2)6-3/h4-11H2,1-3H3 |
| SMILES | CCCCCC(=O)OCCN(CC)CC |
| IUPAC Name | 2-(diethylamino)ethyl hexanoate |
| Molecular Weight (g/mol) | 215.33 |
| Formula Weight | 215.1885 |