missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyguanosine-5'-Triphosphate Trisodium Salt, 98%, MP Biomedicals™

Product Code. 11437670 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11437670

Brand: MP Biomedicals undefined

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 93919-41-6
Assay Percent Range 98%
Molecular Formula C10H13N5Na3O13P3
MDL Number MFCD00080337
Quantity 1 g
Synonym dGTP, 5'-dGMP disodium salt
InChI Key IWGGLKOTEOCWQP-QAUMGBFTNA-K
SMILES [Na+].[Na+].[Na+].NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](COP([O-])(=O)OP([O-])(=O)OP(O)([O-])=O)O2)C(=O)N1
IUPAC Name trisodium ({[(2R,3S,5R)-5-(2-amino-6-oxo-6,9-dihydro-1H-purin-9-yl)-3-hydroxyoxolan-2-yl]methyl phosphonato}oxy)(hydrogen phosphonatooxy)phosphinate
Molecular Weight (g/mol) 573.13
Formula Weight 573
Physical Form Solid
Chemical Name or Material 2'-Deoxyguanosine-5'-Triphosphate Trisodium Salt
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.