missing translation for 'onlineSavingsMsg'
Learn More

2-Deoxy-D-Galactose, MP Biomedicals™

Product Code. 11432202 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 11432202

Brand: MP Biomedicals 02150800.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibitor of fucosylation. 2-Deoxy-D-Galactose, MP Biomedicals is a hexose analog that can be incorporated into rat gangliosides GM2 and GD3 in positions normally occupied by galactose.

  • Hexose analog that can be incorporated into rat gangliosides GM2 and GD3 in positions normally occupied by galactose
  • Has been suggested to act as an inhibitor of fucosylation
  • Also used for competitive elution of Anadarin P lectin (a galactosyl-binding lectin from blood clam)
TRUSTED_SUSTAINABILITY

Specifications

CAS 1949-89-9
Melting Point 109°C to 119°C
Molecular Formula C6H12O5
MDL Number MFCD00014649
Quantity 500 mg
Synonym 2-deoxy-d-galactose, 3r,4r,5r-3,4,5,6-tetrahydroxyhexanal, d-lyxo-hexose, 2-deoxy, unii-531k2iok5q, 2-deoxy-d-lyxo-hexose, lyxo-hexose, 2-deoxy, ksc179c4l
InChI Key PMMURAAUARKVCB-ONLQHTGVNA-N
Optical Rotation 54.8°–66.3° (c=4, H2O)
SMILES OC[C@H]1OC(O)C[C@@H](O)[C@H]1O
IUPAC Name (4R,5R,6R)-6-(hydroxymethyl)oxane-2,4,5-triol
Molecular Weight (g/mol) 164.16
PubChem CID 102191
ChEBI CHEBI:27411
Formula Weight 164.2
Physical Form Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.