missing translation for 'onlineSavingsMsg'
Learn More

2-deoxy-D-galactopyranose, MP Biomedicals™

Product Code. 11462202 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462202 25 g 25g
11452202 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11462202 Supplier MP Biomedicals Supplier No. 0215080025
  • ONLINE EXCLUSIVE PRICE
    17620.00 NOK
    16781.87 NOK / 25g
    Save 838.13 NOK
    4% Off
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1949-89-9
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00014649
InChI Key PMMURAAUARKVCB-ONLQHTGVNA-N
Synonym 2-deoxy-d-galactose, 3r,4r,5r-3,4,5,6-tetrahydroxyhexanal, d-lyxo-hexose, 2-deoxy, unii-531k2iok5q, 2-deoxy-d-lyxo-hexose, lyxo-hexose, 2-deoxy, ksc179c4l
PubChem CID 102191
ChEBI CHEBI:27411
SMILES OC[C@H]1OC(O)C[C@@H](O)[C@H]1O

Specifications

Color White
Assay Percent Range 0.996
Quantity 25 g
IUPAC Name (4R,5R,6R)-6-(hydroxymethyl)oxane-2,4,5-triol
Formula Weight 164.2
Grade Research
Physical Form Powder
Chemical Name or Material 2-Deoxy-D-galactopyranose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.