Learn More
2-Cyano-6-methoxybenzothiazole, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Cyano-6-methoxybenzothiazole |
| CAS | 943-03-3 |
| Molecular Formula | C9 H8 N6 |
| Quantity | 5 g |
| Synonym | 6-Methoxy-2-benzothiazolecarbonitrile, 6-Methoxybenzo[d]thiazole-2-carbonitrile, NSC 377382 |
| InChI Formula | InChI=1S/C9H8N6/c10-12-9-14-13-8-7-4-2-1-3-6(7)5-11-15(8)9/h1-5H,10H2,(H,12,14) |
| SMILES | N\N=C\1/NN=C2N1N=Cc3ccccc23 |
| IUPAC Name | (E)-2H-[1,2,4]triazolo[3,4-a]phthalazin-3-ylidenehydrazine |
| Molecular Weight (g/mol) | 200.2 |
| Formula Weight | 200.081 |