Learn More
2-Cyano-3-methylpyridine, 98%, Thermo Scientific Chemicals
Catalog Number 11378766
Click to view available options
Quantity:
5 g
25 g
100 g
20970-75-6
C7H6N2
118.139
MFCD00190585
WBXZCDIZXWDPBL-UHFFFAOYSA-N
2-cyano-3-methylpyridine, 3-methylpicolinonitrile, 2-cyano-3-methyl pyridine, 3-methyl-2-pyridinecarbonitrile, pyridinecarbonitrile, methyl, 3-methyl-pyridine-2-carbonitrile, 2-cyano-3-methyl-pyridine, 2-pyridinecarbonitrile, 3-methyl, 2-cyano-3-methylpyridine 2-cyano-3-picoline, 2-cyano-3-picoline
819928
3-methylpyridine-2-carbonitrile
CC1=C(N=CC=C1)C#N
20970-75-6
C7H6N2
118.139
MFCD00190585
WBXZCDIZXWDPBL-UHFFFAOYSA-N
2-cyano-3-methylpyridine, 3-methylpicolinonitrile, 2-cyano-3-methyl pyridine, 3-methyl-2-pyridinecarbonitrile, pyridinecarbonitrile, methyl, 3-methyl-pyridine-2-carbonitrile, 2-cyano-3-methyl-pyridine, 2-pyridinecarbonitrile, 3-methyl, 2-cyano-3-methylpyridine 2-cyano-3-picoline, 2-cyano-3-picoline
819928
3-methylpyridine-2-carbonitrile
CC1=C(N=CC=C1)C#N
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 84°C to 87°C
Boiling Point 141°C (38 mmHg)
Flash Point 115°C (239°F)
Odor Characteristic
Quantity 5 g
Beilstein 112001
Formula Weight 118.14
Percent Purity 98%
Chemical Name or Material 2-Cyano-3-methylpyridine