Learn More
Cytiva 2 CHR Cellulose Blotting Paper
Smooth surface. Particularly recommended for optical or radiometric scanning
Brand: Cytiva 3002-911
5700.00 NOK valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 3.56000kg
Description
Thickness 0.18mm. Flow rate 115mm/30 min. Slower than 1 Chr for higher resolution applications. Smooth surface. Particularly recommended for optical or radiometric scanning.- Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind.
- Manufactured and tested specifically for chromatographic techniques. This ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
- Also widely used in protein and nucleic acid blotting.
Specifications
58 x 60 cm (22.8 x 23.6 in.) | |
Grade 2 Chr cellulose chromatography sheets is a 0.18 mm paper for higher resolution applications. | |
Grade 2 Chr | |
Cellulose |