missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorothionicotinamide, 97%, Thermo Scientific™

Catalog Number 15498867
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1240596-59-1
C6H5ClN2S
172.63
MFCD16659867
UIBDAMGTUPCTOT-UHFFFAOYSA-N
2-chlorothionicotinamide, 2-chloro-thionicotinamide, 2-chloropyridine-3-thiocarboxamide, 3-pyridinecarbothioamide, 2-chloro
15631431
2-chloropyridine-3-carbothioamide
C1=CC(=C(N=C1)Cl)C(=S)N
This item is not returnable. View return policy
1240596-59-1
C6H5ClN2S
172.63
MFCD16659867
UIBDAMGTUPCTOT-UHFFFAOYSA-N
2-chlorothionicotinamide, 2-chloro-thionicotinamide, 2-chloropyridine-3-thiocarboxamide, 3-pyridinecarbothioamide, 2-chloro
15631431
2-chloropyridine-3-carbothioamide
C1=CC(=C(N=C1)Cl)C(=S)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 172.63
Percent Purity 97%
Chemical Name or Material 2-Chlorothionicotinamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'