Learn More
2-Chlorophenylacetylene, 98%
Product Code. p-7035715
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
15410258 1 g 1g
15420258 5 g 5g
15430258 25 g 25g
3 options
Product Code. 15410258
Brand: Thermo Scientific Alfa Aesar H53441.03
Request Bulk or Custom Format
CAS: 873-31-4 | C8H5Cl | 136.578 g/mol
2-Chlorophenylacetylene is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chlorophenylacetylene is used as pharmaceutical intermediate.
Solubility
Soluble in hexane.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 873-31-4
Molecular Formula C8H5Cl
Molecular Weight (g/mol) 136.578
MDL Number MFCD00269951
InChI Key DGLHLIWXYSGYBI-UHFFFAOYSA-N
Synonym 2-chlorophenylacetylene, 2'-chlorophenyl acetylene, 1-chloro-2-ethynyl-benzene, benzene, 1-chloro-2-ethynyl, 1-chlor-2-ethinylbenzol, o-chlorphenylacetylen, o-chlorophenylacetylene, pubchem2550, 2-chlorophenyl acetylene, 2;-chlorophenylacetylene
PubChem CID 585995
IUPAC Name 1-chloro-2-ethynylbenzene
SMILES C#CC1=CC=CC=C1Cl
Specifications
Density 1.125
Boiling Point 82°C (12 mmHg)
Quantity 1 g
Refractive Index 1.571
Solubility Information Soluble in hexane.
Formula Weight 136.58
Percent Purity 98%
Chemical Name or Material 2-Chlorophenylacetylene