Learn More
2-Chloroindole-3-carboxaldehyde, 97%
Product Code. 15437317
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15437317 1 g 1g
15447317 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15437317 Supplier Thermo Scientific Alfa Aesar Supplier No. H34381.03
Request Bulk or Custom Format
CAS: 5059-30-3 | C9H6ClNO | 179.603 g/mol
It is used as pharmaceutical intermediates. It is used in the synthesis of indole phytoalexin cyclobrassinon. And also in unprecedented chemical structure and biomimetic synthesis of erucalexin, a phytoalexin from the wild crucifer Erucastrum gallicum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediates. It is used in the synthesis of indole phytoalexin cyclobrassinon. And also in unprecedented chemical structure and biomimetic synthesis of erucalexin, a phytoalexin from the wild crucifer Erucastrum gallicum.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store at room temperature.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5059-30-3
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.603
MDL Number MFCD00111620
InChI Key XYSSNBNFOBVMAU-UHFFFAOYSA-N
Synonym 2-chloro-indole-3-carbaldehyde, 2-chloroindole-3-carbaldehyde, 2-chloro-1h-indole-3-carboxaldehyde, 2-chloroindole-3-carboxaldehyde, 2-chloro-3-formyl-1h-indole, 1h-indole-3-carboxaldehyde, 2-chloro, chloroindolecarbaldehyde, 2-chloro-3-formylindol, 1h-indole-3-carboxaldehyde,2-chloro
PubChem CID 314791
IUPAC Name 2-chloro-1H-indole-3-carbaldehyde
SMILES C1=CC=C2C(=C1)C(=C(N2)Cl)C=O
Specifications
Melting Point ∽212°C (decomposition)
Quantity 1 g
Sensitivity Air sensitive
Solubility Information Slightly soluble in water.
Formula Weight 179.61
Percent Purity 97%
Chemical Name or Material 2-Chloroindole-3-carboxaldehyde