Learn More
2-Chloroimidazole, 97%, Thermo Scientific Chemicals
Catalog Number p-7034196
Click to view available options
Quantity:
250 mg
1 g
5 g
16265-04-6
C3H3ClN2
102.52
MFCD02179530
OCVXSFKKWXMYPF-UHFFFAOYSA-N
2-chloroimidazole, chloroimidazole, sftheadililuh@, pubchem7597, 2-chlor-1h-imidazol, zlchem 1064, acmc-209dos, 1h-imidazole,2-chloro, 2-chloranyl-1h-imidazole, 1h-imidazole, 2-chloro
2773328
CHEBI:52721
2-chloro-1H-imidazole
ClC1=NC=CN1
16265-04-6
C3H3ClN2
102.52
MFCD02179530
OCVXSFKKWXMYPF-UHFFFAOYSA-N
2-chloroimidazole, chloroimidazole, sftheadililuh@, pubchem7597, 2-chlor-1h-imidazol, zlchem 1064, acmc-209dos, 1h-imidazole,2-chloro, 2-chloranyl-1h-imidazole, 1h-imidazole, 2-chloro
2773328
CHEBI:52721
2-chloro-1H-imidazole
ClC1=NC=CN1
2-Chloroimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloroimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 165°C to 166°C
Quantity 250 mg
Formula Weight 102.52
Percent Purity 97%
Chemical Name or Material 2-Chloroimidazole