Learn More
2-Chlorobenzhydrazide, TRC
Product Code. 30357090
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30357090 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30357090 Supplier Toronto Research Chemicals Supplier No. TRCC427643500MG
 Restricted Item
Estimated Shipment: 26-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Chlorobenzhydrazide
CAS 5814-05-1
Molecular Formula C7H7ClN2O
Quantity 500 mg
InChI Formula InChI=1S/C7H7ClN2O/c8-6-4-2-1-3-5(6)7(11)10-9/h1-4H,9H2,(H,10,11)
SMILES NNC(=O)c1ccccc1Cl
IUPAC Name 2-chlorobenzohydrazide
Molecular Weight (g/mol) 170.6
Formula Weight 170.0247