Learn More
2-Chlorobenzhydrazide, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Chlorobenzhydrazide |
| CAS | 5814-05-1 |
| Molecular Formula | C7H7ClN2O |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C7H7ClN2O/c8-6-4-2-1-3-5(6)7(11)10-9/h1-4H,9H2,(H,10,11) |
| SMILES | NNC(=O)c1ccccc1Cl |
| IUPAC Name | 2-chlorobenzohydrazide |
| Molecular Weight (g/mol) | 170.6 |
| Formula Weight | 170.0247 |