Learn More
2-Chlorobenzhydrazide, 98+%
Product Code. 11399639
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399639 5 g 5g
11309649 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11399639 Supplier Thermo Scientific Alfa Aesar Supplier No. A11361.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 5814-05-1 | C7H7ClN2O | 170.60 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5814-05-1
Molecular Formula C7H7ClN2O
Molecular Weight (g/mol) 170.60
MDL Number MFCD00007597
InChI Key KPPNLSKVTKSSTG-UHFFFAOYSA-N
Synonym 2-chlorobenzhydrazide, 2-chlorobenzoic hydrazide, 2-chlorobenzoic acid hydrazide, 2-chlorobenzene-1-carbohydrazide, benzoylhydrazine, o-chloro, benzoic acid, 2-chloro-, hydrazide, 2-chlorobenzenecarbohydrazide, chlorobenzohydrazide, o-chlorobenzhydrazide, 2clphcon2
PubChem CID 79883
IUPAC Name 2-chlorobenzohydrazide
SMILES C1=CC=C(C(=C1)C(=O)NN)Cl
Specifications
Melting Point 116°C to 120°C
Quantity 5 g
Beilstein 972229
Formula Weight 170.6
Percent Purity ≥98%
Chemical Name or Material 2-Chlorobenzhydrazide