Learn More
2-Chlorobenzenesulfonyl chloride, 98%
Product Code. 11457966
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457966 5 g 5g
11467966 25 g 25g
11477966 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11457966 Supplier Thermo Scientific Alfa Aesar Supplier No. A14054.06
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
CAS: 2905-23-9 | C6H4Cl2O2S | 211.06 g/mol
2-Chlorobenzenesulfonyl chloride is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chlorobenzenesulfonyl chloride is used as an organic chemical synthesis intermediate.
Solubility
It reacts with water.
Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2905-23-9
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.06
MDL Number MFCD00051520
InChI Key KMVZDSQHLDGKGV-UHFFFAOYSA-N
Synonym 2-chlorobenzenesulphonyl chloride, 2-chlorobenzene-1-sulfonyl chloride, 2-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 2-chloro, o-chlorobenzenesulfonyl chloride, o-chlorobenzenesulfochloride, 2-chlorophenylsulfonyl chloride, 2-chloro-benzenesulfonyl chloride, benzenesulfonyl chloride, chloro, chloro 2-chlorophenyl sulfone
PubChem CID 76187
IUPAC Name 2-chlorobenzenesulfonyl chloride
SMILES ClC1=CC=CC=C1S(Cl)(=O)=O
Specifications
Melting Point 26°C to 28°C
Density 1.548
Boiling Point 150°C to 152°C (12 mmHg)
Flash Point 140°C (284°F)
Refractive Index 1.576
Quantity 5 g
UN Number UN3265
Beilstein 974321
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 211.07
Percent Purity 98%
Chemical Name or Material 2-Chlorobenzenesulfonyl chloride