Learn More
2′-Chloroacetophenone, Sigma-Aldrich™
Specifications
Specifications
| CAS | 2142-68-9 |
| Density | 1.189 g/cm3 at 20°C |
| MDL Number | MFCD00000560 |
| Quantity | 50 mL |
| Synonym | 2′-Chloroacetophenone, 1-(2-Chlorophenyl)ethanone, 2′-Chloroacetophenone |
| Molecular Weight (g/mol) | 154.59 |
| Grade | Research |