Learn More
BRAND™ Spring Clip
Product Code. p-7164801 Shop All Brand Products
Change view
Click to view available options
Diameter (Metric):
32 mm
51 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10097390 32 mm Each
10488372 51 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10097390 Supplier BRAND™ Supplier No. 76045
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Spring Clips designed for wall mounting of cylindrical apparatus components.
  • Spring steel, coated with zinc and polyethylene
  • Non-scratching
  • Good chemical resistance
  • For wall mounting of cylindrical apparatus components like pipes, tubing, etc.
TRUSTED_SUSTAINABILITY
Specifications
Type Spring Clip
Material Zinc, Steel
Includes 1 Bore, 4 mm
Diameter (Metric) 32 mm
Coating Polyethylene