missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-nitrobenzonitrile, 98%

Product Code. p-7020667
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11430623 5 g 5g
11440623 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11430623

Brand: Thermo Scientific Alfa Aesar A12703.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6575-07-1 | C8H6ClN | 151.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6575-07-1
MDL Number MFCD00007203

Specifications

Melting Point 118°C to 124°C
Molecular Formula C8H6ClN
Quantity 5 g
Beilstein 1369781
Synonym benzonitrile, 2-chloro-6-methyl, 2-cyano-3-chlorotoluene, 3-chloro-2-toluonitrile, o-tolunitrile, 6-chloro, 6-chloro-o-tolunitrile, unii-41m65jiw54, 2-chloro-6-methyl benzonitrile, 2-chloro-6-methylbenzenecarbonitrile, 2-cyano3-chlorotoluene, pubchem23832
InChI Key WQWQHJNUHQEGTN-UHFFFAOYSA-N
SMILES CC1=C(C(=CC=C1)Cl)C#N
IUPAC Name 2-chloro-6-methylbenzonitrile
Molecular Weight (g/mol) 151.59
PubChem CID 23053
Formula Weight 182.57
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-nitrobenzonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.