Learn More
2-Chloro-6-methylbenzaldehyde, 98%
Product Code. 11311949
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11311949 5 g 5g
11301949 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11311949 Supplier Thermo Scientific Alfa Aesar Supplier No. H26023.06
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
CAS: 1194-64-5 | C8H7ClO | 154.593 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1194-64-5
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.593
MDL Number MFCD01934409
InChI Key CCYFXIJPJFSTSU-UHFFFAOYSA-N
PubChem CID 12467050
IUPAC Name 2-chloro-6-methylbenzaldehyde
SMILES CC1=C(C(=CC=C1)Cl)C=O
Specifications
Melting Point 36°C to 38°C
Boiling Point 74°C (0.4 mmHg)
Flash Point 99°C (210°F)
Quantity 5 g
Sensitivity Air Sensitive
Synonym 3-chloro tolualdehyde, pubchem19866, 3-chloro-2-formyltoluene, 2-chloro-6-methylbenzadehyde, 6-chloro-2-methylbenzaldehyde, 2-chloro-6-methyl-benzaldehyde, benzaldehyde,2-chloro-6-methyl, 2-chloranyl-6-methyl-benzaldehyde, benzaldehyde, 2-chloro-6-methyl, 2-chloro-6-methylbenzaldehyde
Formula Weight 154.59
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-methylbenzaldehyde