Learn More
2-Chloro-5-methylaniline, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-5-methylaniline |
| CAS | 95-81-8 |
| Molecular Formula | C7 H8 Cl N |
| Quantity | 1 g |
| Synonym | m-Toluidine, 6-chloro- (8CI), 2-Chloro-5-methylbenzenamine, 2-Chloro-5-methylaniline, 3-Amino-4-chlorotoluene, 6-Chloro-m-toluidine |
| InChI Formula | InChI=1S/C7H8ClN/c1-5-2-3-6(8)7(9)4-5/h2-4H,9H2,1H3 |
| SMILES | Cc1ccc(Cl)c(N)c1 |
| IUPAC Name | 2-chloro-5-methylaniline |
| Molecular Weight (g/mol) | 141.6 |
| Formula Weight | 141.0345 |