missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluorobenzoyl chloride, 97%, Thermo Scientific™

Catalog Number 11490990
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
21900-51-6
C7H3Cl2FO
192.998
MFCD01631417
SGZSJPBASHYOHQ-UHFFFAOYSA-N
2-chloro-5-fluorobenzoylchloride, 5-fluoro-2-chlorobenzoyl chloride, benzoyl chloride, 2-chloro-5-fluoro, acmc-1cg8v, attercop-chm at108545, 2-chloro-5-fluoro-benzoyl chloride, benzoyl chloride,2-chloro-5-fluoro, 2-chlorocarbonyl-4-fluorochlorobenzene, 2-chloranyl-5-fluoranyl-benzoyl chloride, benzoyl chloride, 2-chloro-5-fluoro-8ci,9ci
2757540
2-chloro-5-fluorobenzoyl chloride
C1=CC(=C(C=C1F)C(=O)Cl)Cl
This item is not returnable. View return policy
21900-51-6
C7H3Cl2FO
192.998
MFCD01631417
SGZSJPBASHYOHQ-UHFFFAOYSA-N
2-chloro-5-fluorobenzoylchloride, 5-fluoro-2-chlorobenzoyl chloride, benzoyl chloride, 2-chloro-5-fluoro, acmc-1cg8v, attercop-chm at108545, 2-chloro-5-fluoro-benzoyl chloride, benzoyl chloride,2-chloro-5-fluoro, 2-chlorocarbonyl-4-fluorochlorobenzene, 2-chloranyl-5-fluoranyl-benzoyl chloride, benzoyl chloride, 2-chloro-5-fluoro-8ci,9ci
2757540
2-chloro-5-fluorobenzoyl chloride
C1=CC(=C(C=C1F)C(=O)Cl)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 106°C (18 mmHg)
Refractive Index 1.55
Quantity 1 g
UN Number UN3265
Beilstein 2640754
Sensitivity Moisture sensitive
Formula Weight 193.01
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-fluorobenzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'