Learn More
2-Chloro-4-(methylcarbamoyl)benzeneboronic acid, 97%
Product Code. 15412286
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15412286 1 g 1g
15422286 250 mg 250mg
2 options
Product Code. 15412286
Brand: Thermo Scientific Alfa Aesar H55778.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 1451391-89-1 | C8H9BClNO3 | 213.424 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1451391-89-1
Molecular Formula C8H9BClNO3
Molecular Weight (g/mol) 213.424
MDL Number MFCD22125174
InChI Key LSMHGCJWZPHACP-UHFFFAOYSA-N
Synonym 2-chloro-4-methylcarbamoyl phenylboronic acid, 2-chloro-4-methylcarbamoyl benzeneboronic acid, 2-chloro-4-methylcarbamoyl phenyl boronic acid
PubChem CID 73996177
IUPAC Name [2-chloro-4-(methylcarbamoyl)phenyl]boronic acid
SMILES B(C1=C(C=C(C=C1)C(=O)NC)Cl)(O)O
Specifications
Quantity 1 g
Formula Weight 213.43
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-(methylcarbamoyl)benzeneboronic acid