missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-hydroxybenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number 15472118
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
56962-11-9
C7H5ClO2
156.565
MFCD00052184
ZMOMCILMBYEGLD-UHFFFAOYSA-N
benzaldehyde, 2-chloro-4-hydroxy, 2-chloro-4-hydroxy-benzaldehyde, benzaldehyde, chloro-4-hydroxy, 2-chloro-4-hydroxybenzaldehyde, pubchem7366, acmc-209lv6, ksc497o6r, 2-chlor-4-hydroxy-benzaldehyd, 2-chloranyl-4-oxidanyl-benzaldehyde
185363
2-chloro-4-hydroxybenzaldehyde
C1=CC(=C(C=C1O)Cl)C=O
This item is not returnable. View return policy
56962-11-9
C7H5ClO2
156.565
MFCD00052184
ZMOMCILMBYEGLD-UHFFFAOYSA-N
benzaldehyde, 2-chloro-4-hydroxy, 2-chloro-4-hydroxy-benzaldehyde, benzaldehyde, chloro-4-hydroxy, 2-chloro-4-hydroxybenzaldehyde, pubchem7366, acmc-209lv6, ksc497o6r, 2-chlor-4-hydroxy-benzaldehyd, 2-chloranyl-4-oxidanyl-benzaldehyde
185363
2-chloro-4-hydroxybenzaldehyde
C1=CC(=C(C=C1O)Cl)C=O

2-Chloro-4-hydroxybenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-4-hydroxybenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in organic solvents.

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 145°C to 147°C
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Soluble in organic solvents.
Formula Weight 156.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-hydroxybenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'