missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,5-difluorobenzonitrile, 97%, Thermo Scientific™

Product Code. 11431200
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431200 10 g 10g
11421200 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11431200 Supplier Thermo Scientific Alfa Aesar Supplier No. H26322.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135748-34-4
Molecular Formula C7H2ClF2N
Molecular Weight (g/mol) 173.55
MDL Number MFCD02871619
InChI Key AKZDICFDUJUQRX-UHFFFAOYSA-N
PubChem CID 4085849
IUPAC Name 2-chloro-4,5-difluorobenzonitrile
SMILES FC1=CC(Cl)=C(C=C1F)C#N

Specifications

Melting Point 33°C to 36°C
Quantity 10 g
UN Number UN3439
Synonym 2-chloro-4,5-difluoro-benzonitrile, benzonitrile, 2-chloro-4,5-difluoro, 2-chloro-4,5-difluorobenzenecarbonitrile, pubchem1567, intermediates-zcf02046, acmc-1cg62, benzonitrile,2-chloro-4,5-difluoro, 2-chloro-4,5-difluorobenzonitrile, 2-chloro-4,5-difluorobenzonitrile 2g
Formula Weight 173.55
Percent Purity 97%
Chemical Name or Material 2-Chloro-4,5-difluorobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.