Learn More
2-Chloro-4,5-difluorobenzonitrile, 97%, Thermo Scientific™
Product Code. 11421200
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11421200 1 g 1g
11431200 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11421200 Supplier Thermo Scientific Alfa Aesar Supplier No. H26322.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 135748-34-4
Molecular Formula C7H2ClF2N
Molecular Weight (g/mol) 173.55
MDL Number MFCD02871619
InChI Key AKZDICFDUJUQRX-UHFFFAOYSA-N
PubChem CID 4085849
IUPAC Name 2-chloro-4,5-difluorobenzonitrile
SMILES FC1=CC(Cl)=C(C=C1F)C#N
Specifications
Melting Point 33°C to 36°C
Quantity 1 g
UN Number UN3439
Synonym 2-chloro-4,5-difluoro-benzonitrile, benzonitrile, 2-chloro-4,5-difluoro, 2-chloro-4,5-difluorobenzenecarbonitrile, pubchem1567, intermediates-zcf02046, acmc-1cg62, benzonitrile,2-chloro-4,5-difluoro, 2-chloro-4,5-difluorobenzonitrile, 2-chloro-4,5-difluorobenzonitrile 2g
Formula Weight 173.55
Percent Purity 97%
Chemical Name or Material 2-Chloro-4,5-difluorobenzonitrile