Learn More
2-Chloro-3-methylquinoline, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Chloro-3-methylquinoline is used as organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
2-Chloro-3-methylquinoline is used as organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | 82°C to 84°C |
Quantity | 1 g |
Solubility Information | Insoluble in water. |
Formula Weight | 177.63 |
Percent Purity | 97% |
Chemical Name or Material | 2-Chloro-3-methylquinoline |