Learn More
2-Chloro-3-iodo-1,4-dimethoxybenzene, 97%
Product Code. 15457167
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15457167 5 g 5g
15447167 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15457167 Supplier Thermo Scientific Alfa Aesar Supplier No. H34232.06
Request Bulk or Custom Format
CAS: 1335243-55-4 | C8H8ClIO2 | 298.504 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1335243-55-4
Molecular Formula C8H8ClIO2
Molecular Weight (g/mol) 298.504
MDL Number MFCD22380712
InChI Key IMMMDYMSUZKGSS-UHFFFAOYSA-N
Synonym 1-chloro-3,6-dimethoxy-2-iodobenzene
PubChem CID 98001136
IUPAC Name 2-chloro-3-iodo-1,4-dimethoxybenzene
SMILES COC1=C(C(=C(C=C1)OC)I)Cl
Specifications
Melting Point 129°C to 131°C
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 298.51
Percent Purity 97%
Chemical Name or Material 2-Chloro-3-iodo-1,4-dimethoxybenzene