Learn More
2-chloro-3-(chloromethyl)quinoline, 97%, Thermo Scientific™
Product Code. p-3075410
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. 10523983
Brand: Thermo Scientific Maybridge MO07048CB
Request Bulk or Custom Format
Chemical Identifiers
CAS 90097-52-2
Molecular Formula C10H7Cl2N
Molecular Weight (g/mol) 212.073
MDL Number MFCD05865134
InChI Key UOIJHUIOTFDGNA-UHFFFAOYSA-N
Synonym 2-chloro-3-chloromethyl quinoline, quinoline, 2-chloro-3-chloromethyl, 2-chloranyl-3-chloromethyl quinoline
PubChem CID 2063384
IUPAC Name 2-chloro-3-(chloromethyl)quinoline
SMILES C1=CC=C2C(=C1)C=C(C(=N2)Cl)CCl
Specifications
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 212.08
Percent Purity 97%
Chemical Name or Material 2-chloro-3-(chloromethyl)quinoline