Learn More
2-Chloro-3',4'-dihydroxyacetophenone, 97%
Codice prodotto. p-7027046
Cambia vista
Click to view available options
Quantity:
25 g
100 g
500 g
Dimensione della confezione:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
11475497 25 g 25g
11485497 100 g 100g
11495497 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11475497 Fornitore Thermo Scientific Alfa Aesar N. del fornitore B23496.14
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
CAS: 99-40-1 | C8H7ClO3 | 186.591 g/mol
2-Chloro-3',4'-dihydroxyacetophenone is used as a styptic adrenobazone, quasi adrenaline drug gasp spirit of intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloro-3′,4′-dihydroxyacetophenone is used as a styptic adrenobazone, quasi adrenaline drug gasp spirit of intermediates.
Solubility
Soluble in DMSO, methanol.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 99-40-1
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00002200
InChI Key LWTJEJCZJFZKEL-UHFFFAOYSA-N
Synonym 2-chloro-3',4'-dihydroxyacetophenone, 2-chloro-1-3,4-dihydroxyphenyl ethanone, 4-chloroacetyl catechol, 3,4-dihydroxyphenacyl chloride, ethanone, 2-chloro-1-3,4-dihydroxyphenyl, unii-fn6949l56q, 3',4'-dihydroxy-2-chloroacetophenone, chloromethyl 3,4-dihydroxyphenyl ketone, 2-chloro-3,4-dihydroxyacetophenone, .alpha.-chloro-3,4-dihydroxyacetophenone
PubChem CID 66834
ChEBI CHEBI:51844
IUPAC Name 2-chloro-1-(3,4-dihydroxyphenyl)ethanone
SMILES C1=CC(=C(C=C1C(=O)CCl)O)O
Specifica
Melting Point 172°C to 176°C
Quantity 25 g
Beilstein 2092660
Solubility Information Soluble in DMSO,methanol.
Formula Weight 186.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-3',4'-dihydroxyacetophenone
compliance-icons
Identificativo prodotto
  • 2-Chloro-3',4'-dihydroxyacetophenone
Avvertenza
  • Warning
Categoria di pericolo
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Consigli di prudenza
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Informazioni supplementari
  • MIXTURE LIST-Contain: 2-Chloro-3',4'-dihydroxyacetophenone
RUO – Research Use Only
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.