Learn More
2-Chloro-1-(2,4-difluorophenyl)ethanone, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 51336-94-8 |
| Molecular Formula | C8H5ClF2O |
| MDL Number | MFCD00013252 |
| Quantity | 25 g |
| SMILES | FC1=CC=C(C(CCl)=O)C(F)=C1 |
| Molecular Weight (g/mol) | 190.57 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Chloro-1-(2,4-difluorophenyl)ethanone |