missing translation for 'onlineSavingsMsg'
Learn More

2-Carboxyphenyl phosphate, 98%

Product Code. 11457047
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11457047

Brand: Thermo Scientific Alfa Aesar A14417.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6064-83-1 | C7H7O6P | 218.101 g/mol

2-Carboxyphenyl phosphate is used as a substrate for the estimation of acid phosphatase. It is also used as a analgesic, anti-inflammatory agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Carboxyphenyl phosphate is used as a substrate for the estimation of acid phosphatase. It is also used as a analgesic, anti-inflammatory agent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6064-83-1
Molecular Formula C7H7O6P
Molecular Weight (g/mol) 218.101
MDL Number MFCD00042645
InChI Key FFKUDWZICMJVPA-UHFFFAOYSA-N
Synonym fosfosal, 2-phosphonooxy benzoic acid, disdolen, salicyl phosphate, o-carboxyphenyl phosphate, 2-carboxyphenyl phosphate, 2-phosphonoxybenzoic acid, phosphonoxybenzoic acid, o-phosphonooxy benzoic acid, fosfosal inn
PubChem CID 3418
IUPAC Name 2-phosphonooxybenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)OP(=O)(O)O

Specifications

Melting Point 162°C to 166°C
Quantity 25 g
Beilstein 2619142
Merck Index 14,4255
Solubility Information Soluble in water.
Formula Weight 218.1
Percent Purity 98%
Chemical Name or Material 2-Carboxyphenyl phosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.