Learn More
2-Butyne-1,4-diol, Sigma-Aldrich™
Product Code. 15838570
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15838570 500 g 500g
15828570 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15838570 Supplier Sigma Aldrich Supplier No. 8015370500
 Restricted Item
Estimated Shipment: 26-11-2026
Not stabilized for synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 110-65-6
Grade Research
Molecular Weight (g/mol) 86.09
Specifications
Melting Point 54°C to 57°C
Density 1.11 g/cm3 at 20°C
Boiling Point 125°C to 127°C/3 hPa
MDL Number MFCD00002915
Quantity 500 g
Synonym 2-Butyne-1,4-diol, 1,4-Dimethoxyacetylene, 2-Butyne-1,4-diol