Learn More
2-Butanol, Supelco™
Product Code. 15821741
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15821741 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15821741 Supplier Supelco Supplier No. 8222632500
 Restricted Item
Estimated Shipment: 23-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
EMPLURA™
TRUSTED_SUSTAINABILITY
Specifications
Melting Point -115°C
CAS 78-92-2
Density 0.81 g/cm3 at 20°C
Quantity 2.5 L
Molecular Formula CH3CH(OH)CH2CH3
MDL Number MFCD00004569
Synonym 2-Butanol, sec-Butyl alcohol, 2-Butanol
Boiling Point 99°C to 102°C/1013 hPa
Molecular Weight (g/mol) 74.12
Grade Research
Chemical Name or Material 2-Butanol