Learn More
2-Bromophenylacetylene, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Bromophenylacetylene |
| CAS | 766-46-1 |
| Molecular Formula | C8 H5 Br |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C8H5Br/c1-2-7-5-3-4-6-8(7)9/h1,3-6H |
| SMILES | Brc1ccccc1C#C |
| IUPAC Name | 1-bromo-2-ethynylbenzene |
| Molecular Weight (g/mol) | 181.03 |
| Formula Weight | 179.9575 |