Learn More
2-Bromoimidazole, 95%
Product Code. 15409297
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15409297
Brand: Thermo Scientific Alfa Aesar H52553.06
Request Bulk or Custom Format
CAS: 16681-56-4 | C3H3BrN2 | 146.98 g/mol
2-Bromoimidazole is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromoimidazole is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 16681-56-4
Molecular Formula C3H3BrN2
Molecular Weight (g/mol) 146.98
MDL Number MFCD02179526
InChI Key AXHRGVJWDJDYPO-UHFFFAOYSA-N
Synonym 2-bromoimidazole, 1h-imidazole, 2-bromo, bromoimidazole, sftheabililuh@, pubchem8987, zlchem 1065, acmc-1btox, 2-bromanyl-1h-imidazole, ksc176e6b, 2-bromo-1h-imidazole
PubChem CID 2773261
IUPAC Name 2-bromo-1H-imidazole
SMILES BrC1=NC=CN1
Specifications
Melting Point 197°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 146.98
Percent Purity 95%
Chemical Name or Material 2-Bromoimidazole