Learn More
2-Bromohexanoic acid, 97%
Product Code. 11441267
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. 11441267
Brand: Thermo Scientific Alfa Aesar B24228.22
Request Bulk or Custom Format
CAS: 616-05-7 | C6H11BrO2 | 195.056 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 616-05-7
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.056
MDL Number MFCD00004218
InChI Key HZTPKMIMXLTOSK-UHFFFAOYSA-N
Synonym 2-bromocaproic acid, dl-2-bromohexanoic acid, hexanoic acid, 2-bromo, alpha-bromohexanoic acid, alpha-bromo-n-caproic acid, .alpha.-bromo-n-caproic acid, 2-bromo-hexanoic acid, .alpha.-bromocaproic acid, .alpha.-bromohexanoic acid, 2-bromo-n-hexanoic acid
PubChem CID 12013
IUPAC Name 2-bromohexanoic acid
SMILES CCCCC(C(=O)O)Br
Specifications
Melting Point 3°C to 5°C
Density 1.37
Boiling Point 136°C to 138°C (18 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.472
Quantity 100 g
UN Number UN3265
Merck Index 14,1415
Formula Weight 195.06
Percent Purity 97%
Chemical Name or Material 2-Bromohexanoic acid