Learn More
2-Bromoethyl Benzoate (~90%), TRC
Specifications
Specifications
| Chemical Name or Material | 2-Bromoethyl Benzoate |
| CAS | 939-54-8 |
| Molecular Formula | C9H9O2Br |
| Quantity | 500 mg |
| Synonym | beta-Bromoethyl Benzoate |
| InChI Formula | InChI=1S/C9H9BrO2/c10-6-7-12-9(11)8-4-2-1-3-5-8/h1-5H,6-7H2 |
| SMILES | BrCCOC(=O)c1ccccc1 |
| IUPAC Name | 2-bromoethyl benzoate |
| Molecular Weight (g/mol) | 229.07 |
| Formula Weight | 227.9786 |