missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromoethoxy)-tert-butyldimethylsilane, 98+%, stabilized, Thermo Scientific Chemicals

Catalog Number p-3771077
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
86864-60-0
C8H19BrOSi
239.22
MFCD00209550
JBKINHFZTVLNEM-UHFFFAOYSA-N
2-bromoethoxy-tert-butyldimethylsilane, 2-bromoethoxy tert-butyl dimethylsilane, 2-bromoethoxy-t-butyl dimethylsilane, 2-t-butyldimethylsiloxy ethylbromide, 2-bromoethoxy-t-butyldimethylsilane, 2-bromo-ethoxy-tert-butyl-dimethyl-silane, silane, 2-bromoethoxy 1,1-dimethylethyl dimethyl, brch2ch2otbs
3608067
2-bromoethoxy-tert-butyl-dimethylsilane
CC(C)(C)[Si](C)(C)OCCBr
This item is not returnable. View return policy
86864-60-0
C8H19BrOSi
239.22
MFCD00209550
JBKINHFZTVLNEM-UHFFFAOYSA-N
2-bromoethoxy-tert-butyldimethylsilane, 2-bromoethoxy tert-butyl dimethylsilane, 2-bromoethoxy-t-butyl dimethylsilane, 2-t-butyldimethylsiloxy ethylbromide, 2-bromoethoxy-t-butyldimethylsilane, 2-bromo-ethoxy-tert-butyl-dimethyl-silane, silane, 2-bromoethoxy 1,1-dimethylethyl dimethyl, brch2ch2otbs
3608067
2-bromoethoxy-tert-butyl-dimethylsilane
CC(C)(C)[Si](C)(C)OCCBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Color White to Yellow
Density 1.1150g/mL
Boiling Point 70°C to 75°C (2.5 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)CSi(CH3)2O(CH2)2Br
Quantity 1 g
Specific Gravity 1.115
Stabilizer 5 % max. Na2CO3
Formula Weight 239.22
Percent Purity 98+%
Physical Form Crystalline Solid or Crystals
Chemical Name or Material (2-Bromoethoxy)-tert-butyldimethylsilane
Show More Show Less
compliance-icons
Product Identifier
  • (2-Bromoethoxy)-tert-butyldimethylsilane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'