missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7021965
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
7154-66-7
C7H4BrClO
219.462
MFCD00000655
NZCKTGCKFJDGFD-UHFFFAOYSA-N
benzoyl chloride, 2-bromo, o-bromobenzoyl chloride, benzoyl chloride, o-bromo, 2-bromo-benzoyl chloride, 2-bromobenzoylchloride, 2-bromobenzene-1-carbonyl chloride, o-brombenzoylchlorid, bromo benzoylchloride, o-bromobenzoylchloride, pubchem12510
23542
2-bromobenzoyl chloride
C1=CC=C(C(=C1)C(=O)Cl)Br
This item is not returnable. View return policy
7154-66-7
C7H4BrClO
219.462
MFCD00000655
NZCKTGCKFJDGFD-UHFFFAOYSA-N
benzoyl chloride, 2-bromo, o-bromobenzoyl chloride, benzoyl chloride, o-bromo, 2-bromo-benzoyl chloride, 2-bromobenzoylchloride, 2-bromobenzene-1-carbonyl chloride, o-brombenzoylchlorid, bromo benzoylchloride, o-bromobenzoylchloride, pubchem12510
23542
2-bromobenzoyl chloride
C1=CC=C(C(=C1)C(=O)Cl)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 10°C
Density 1.674
Boiling Point 245°C
Flash Point 107°C (224°F)
Refractive Index 1.596
Quantity 10 g
UN Number UN3265
Beilstein 508506
Sensitivity Moisture sensitive
Formula Weight 219.47
Percent Purity 98%
Chemical Name or Material 2-Bromobenzoyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'