missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-m-xylene, 98+%

Product Code. 11369659
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
250 g
500 g
Unit Size:
100g
10g
250g
25g
500g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369659 25 g 25g
11404494 10 g 10g
11379659 100 g 100g
11414494 250 g 250g
11389659 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11369659 Supplier Thermo Scientific Alfa Aesar Supplier No. A13130.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 576-22-7 | C8H9Br | 185.064 g/mol

2-Bromo-m-xylene has been used to prepare 2,2,4,6,6-pentamethylbiphenyl. It is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-m-xylene has been used to prepare 2,2′,4,6,6′-pentamethylbiphenyl. It is used as an intermediate in organic synthesis.

Solubility
Soluble in aqueous solution. Very soluble in ethanol, soluble in acetone and benzene.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents. Should be stored in a tightly closed container. Store in a cool, dry, well-ventilated area.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 576-22-7
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000075
InChI Key MYMYVYZLMUEVED-UHFFFAOYSA-N
Synonym 2,6-dimethylbromobenzene, 2-bromo-m-xylene, 1-bromo-2,6-dimethylbenzene, benzene, 2-bromo-1,3-dimethyl, 2,6-dimethyl bromobenzene, m-xylene, 2-bromo, 2,6-dimethylphenyl bromide, 2-bromo-meta-xylene, 2-bromo-1,3-dimethyl-benzene, pubchem3206
PubChem CID 68471
IUPAC Name 2-bromo-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)Br

Specifications

Melting Point -10°C
Density 1.384
Quantity 25 g
Refractive Index 1.555
Beilstein 1929780
Boiling Point 205°C to 206°C
Solubility Information Soluble in aqueous solution. Very soluble in ethanol,soluble in acetone and benzene.
Formula Weight 185.07
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-m-xylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.