missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-fluorobenzotrifluoride, 97%, Thermo Scientific™

Product Code. 11421230
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11421230 1 g 1g
15483447 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 11421230

Brand: Thermo Scientific Alfa Aesar H26337.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261951-85-3
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 242.999
MDL Number MFCD01631569
InChI Key LOXSSZUHCPBDMD-UHFFFAOYSA-N
Synonym 2-bromo-6-fluorobenzotrifluoride, 1-bromo-3-fluoro-2-trifluoromethyl benzene, 3-fluoro-2-trifluoromethyl bromobenzene, benzene, 1-bromo-3-fluoro-2-trifluoromethyl, 2-bromo-alpha,alpha,alpha,6-tetrafluorotoluene, 1-bromo-3-fluoro-2-trifluoromethyl-benzene, pubchem1638, 2-bromo-6-fluorobenzo trifluoride
PubChem CID 2773345
IUPAC Name 1-bromo-3-fluoro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C(=C1)Br)C(F)(F)F)F

Specifications

Refractive Index 1.472
Quantity 1 g
Formula Weight 243
Percent Purity 97%
Chemical Name or Material 2-Bromo-6-fluorobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.