missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluorobenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog Number 11447076
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
394-28-5
C7H3BrFO2
218.00
MFCD00142874
OQBMJMJZMDBQSM-UHFFFAOYSA-M
benzoic acid, 2-bromo-5-fluoro, 5-fluoro-2-bromobenzoic acid, 2-bromo-5-fluoro-benzoic acid, 2-bromo-5-fluorobenzoicacid, 2-bromo-5-fluorbenzoic acid, buttpark 19\01-66, pubchem1315, acmc-1ahhq, ksc226g3p, rarechem al bo 0747
2778181
2-bromo-5-fluorobenzoic acid
[O-]C(=O)C1=CC(F)=CC=C1Br
This item is not returnable. View return policy
394-28-5
C7H3BrFO2
218.00
MFCD00142874
OQBMJMJZMDBQSM-UHFFFAOYSA-M
benzoic acid, 2-bromo-5-fluoro, 5-fluoro-2-bromobenzoic acid, 2-bromo-5-fluoro-benzoic acid, 2-bromo-5-fluorobenzoicacid, 2-bromo-5-fluorbenzoic acid, buttpark 19\01-66, pubchem1315, acmc-1ahhq, ksc226g3p, rarechem al bo 0747
2778181
2-bromo-5-fluorobenzoic acid
[O-]C(=O)C1=CC(F)=CC=C1Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 154°C to 158°C
Quantity 1 g
Beilstein 2575978
Formula Weight 219.01
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-5-fluorobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'