Learn More
2-Bromo-4-fluoro-6-(trifluoromethyl)aniline, 98%
Product Code. 11326629
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326629 1 g 1g
11336629 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11326629 Supplier Thermo Scientific Alfa Aesar Supplier No. H26206.03
Request Bulk or Custom Format
CAS: 875664-27-0 | C7H4BrF4N | 258.014 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 875664-27-0
Molecular Formula C7H4BrF4N
Molecular Weight (g/mol) 258.014
MDL Number MFCD04973756
InChI Key OTYCOVZULSPPPW-UHFFFAOYSA-N
Synonym 2-bromo-4-fluoro-6-trifluoromethyl aniline, 2-amino-3-bromo-5-fluorobenzotrifluoride, buttpark 154\11-38, benzenamine,2-bromo-4-fluoro-6-trifluoromethyl, 2-bromo-4-fluoro-6-trifluoromethyl benzenamine, pubchem19869, ksc658m8n, 2-bromo-4-fluoro-6-trifluoromethyl phenylamine, 6-bromo-4-fluoro-2-trifluoromethyl phenylamine, 2-bromanyl-4-fluoranyl-6-trifluoromethyl aniline
PubChem CID 7018045
IUPAC Name 2-bromo-4-fluoro-6-(trifluoromethyl)aniline
SMILES C1=C(C=C(C(=C1Br)N)C(F)(F)F)F
Specifications
Quantity 1 g
UN Number UN2810
Formula Weight 258.01
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-fluoro-6-(trifluoromethyl)aniline